Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

424M-1

Sigma-Aldrich

CITED1 (5H6) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$273.00
0.1 ML
$297.00
7 ML
$808.00
1 ML
$1,210.00

$273.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$273.00
0.1 ML
$297.00
7 ML
$808.00
1 ML
$1,210.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$273.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

5H6, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (424M-14)
vial of 0.5 mL concentrate (424M-15)
bottle of 1.0 mL predilute (424M-17)
vial of 1.0 mL concentrate (424M-16)
bottle of 7.0 mL predilute (424M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG2aκ

control

Wilms’ tumor

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... CITED1(4435)

General description

CITED1 is a transcriptional cofactor expressed in the metanephric mesenchyme (MM) of the embryonic kidney and is down-regulated as these cells undergo epithelial differentiation.1-4 It is thought that CITED1 may play a role in maintaining MM cells in an undifferentiated state.3 Wilms′ tumors (WT) are thought to arise from abnormal postnatal retention and dysregulated differentiation of nephrogenic progenitor cells that originate as a condensed MM within embryonic kidneys.1 CITED1 expression has been shown to persist in blastemal cell populations of human WT.1,2 In the developing embryonic kidney, CITED1 expression is seen in the cytoplasmic compartment. In WT, expression of CITED1 is detected in the nuclear compartment of tumor cells.1-4 It has been suggested that persistent expression of CITED1 in the MM could play a role in WT initiation and pathogenesis.1 CITED1 has been detected in 86.8% of WT Cases.2

Quality


IVD

IVD

IVD

RUO

Linkage

CITED1 Positive Control Slides, Product No. 424S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harold N Lovvorn et al.
Neoplasia (New York, N.Y.), 9(7), 589-600 (2007-08-22)
Wilms' tumors, or nephroblastomas, are thought to arise from abnormal postnatal retention and dysregulated differentiation of nephrogenic progenitor cells that originate as a condensed metanephric mesenchyme within embryonic kidneys. We have previously shown that the transcriptional regulator CITED1 (CBP/p300-interacting transactivators
Scott Boyle et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 236(8), 2321-2330 (2007-07-07)
Early kidney development in mammals is characterized by reciprocal tissue interaction between the ureteric bud and the metanephric mesenchyme. The coordinated response to this interaction is regulated largely at the transcriptional level. Here, we investigate the expression and function of
Andrew J Murphy et al.
Journal of pediatric surgery, 47(6), 1239-1249 (2012-06-19)
SIX2 and CITED1 are transcriptional regulators that specify self-renewing nephronic progenitor cells of the embryonic kidney. We hypothesized that SIX2, which promotes and maintains this stem cell population, and CITED1 remain active in Wilms' tumor (WT). To evaluate expression domains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service