Skip to Content
MilliporeSigma
All Photos(1)

Documents

41629

Sigma-Aldrich

LUCY® 569 solution

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12171500

λmax

~559 nm in methanol

fluorescence

λex 565 nm; λem 581 nm in aqueous buffer pH 8.3 with SDS and BSA

storage temp.

2-8°C

Application

LUCY 569 is a fluorescent stain for protein electrophoresis, with good sensitivity and easy, fast and robust staining procedure for all kinds of SDS gels. LUCY 569 provides a broad linear dynamic range for protein quantification in electrophoresis. Protein staining by LUCY 569 does not interfere with subsequent MALDI-MS.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Physical form

LUCY 569 is provided as a 5000× stock solution in DMSO (5 mg/ml).

Analysis Note

Sensitivity of routine procedure is ca. 5 ng/band, with linear response between 5 and 6000 ng/band.

Legal Information

LUCY is a registered trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service