Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

41367

Sigma-Aldrich

Anti-Rabbit IgG−Abberior® STAR 580 antibody produced in goat

Synonym(s):

Abberior® STAR 580-Anti-Rabbit IgG antibody produced in goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.45

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rabbit

concentration

~1 mg/mL

fluorescence

λex 587 nm; λem 607 nm in PBS, pH 7.4

storage temp.

−20°C

General description

Abberior STAR 580 was developed for STED and confocal microscopy in the orange spectral region. Abberior STAR 580 is highly photostable and bright orange fluorescent dye. The dye works extremely well with Abberior Instruments STEDYCON and Expert Line Microscopes and Leica STED microscopes. It can be very effectively excited with excitation wavelength between 550 - 600 nm. Abberior STAR 580 can substitute dyes like Cy3, Atto 590 or Alexa Fluor 568. For STED microscopy, Abberior STAR 580 can be used most efficiently with a STED Laser wavelength between 750 - 800 nm. Abberior STAR 580 is the ideal partner for Abberior STAR RED or Abberior STAR 635P to obtain optimal 2 Color STED results. Best results are obtained with freshly prepared samples.


Photophysical properties (carboxylic acid):
Absorption Maximum, λex [nm]: 587 (PBS pH 7.4), 584 (H2O), 584 (MeOH)
Extinction Coefficient, εmax [M-1cm-1]: 85 000 (PBS pH 7.4), 85 000 (H2O), 95 000 (MeOH)
Correction Factor, CF260 = ε260max: 0,17 (PBS pH 7.4)
Correction Factor, CF280 = ε280max: 0,17 (PBS pH 7.4)
Fluorescence Maximum, λem [nm]: 607 (PBS pH 7.4), 607 (H2O), 604 (MeOH)
Recommended STED Wavelength, λ [nm]: 750-800
Fluorescence Quantum Yield, λ: 0,90 (PBS pH 7.4)
Fluorescence Lifetime, τ [ns]: 3,5 (PBS pH 7.4)

Suitability

Designed and tested for fluorescent super-resolution microscopy

Analysis Note

May contain significant amounts of BSA as stabilizer
unconjugated dye ≤5% of total fluorescence

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
Atto is a trademark of Atto-Tec GmbH
Cy3 is a trademark of Cytiva
abberior is a registered trademark of Abberior GmbH

Not finding the right product?  

Try our Product Selector Tool.

related product

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eulashini Chuntharpursat-Bon et al.
Communications biology, 6(1), 358-358 (2023-04-04)
Two prominent concepts for the sensing of shear stress by endothelium are the PIEZO1 channel as a mediator of mechanically activated calcium ion entry and the PECAM1 cell adhesion molecule as the apex of a triad with CDH5 and VGFR2.
Francisco Balzarotti et al.
ACS nano, 6(6), 4580-4584 (2012-06-19)
Plasmonics and near-field optical nanoscopy both deal with expanding optics into the subwavelength regime. However, these two fields have so far followed parallel paths of development and only recently have researchers started to explore combinations of their concepts with potential
Esley Torres-García et al.
Nature communications, 13(1), 7452-7452 (2022-12-03)
The resolution of fluorescence microscopy images is limited by the physical properties of light. In the last decade, numerous super-resolution microscopy (SRM) approaches have been proposed to deal with such hindrance. Here we present Mean-Shift Super Resolution (MSSR), a new
Femmy C Stempels et al.
The Journal of cell biology, 222(7) (2023-05-18)
Endosomal Sorting Complex Required for Transport (ESCRT) proteins can be transiently recruited to the plasma membrane for membrane repair and formation of extracellular vesicles. Here, we discovered micrometer-sized worm-shaped ESCRT structures that stably persist for multiple hours at the plasma
Aleksandar Salim et al.
JACS Au, 3(8), 2247-2256 (2023-09-01)
Polo-like kinase (Plk4) is a serine/threonine-protein kinase that is essential for biogenesis of the centriole organelle and is enriched at centrioles. Herein, we introduce Cen-TCO, a chemical probe based on the Plk4 inhibitor centrinone, to image Plk4 and centrioles in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service