Skip to Content
MilliporeSigma
All Photos(1)

Documents

41087

Sigma-Aldrich

Fluorescent Red Mega 480

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30N2O7S
CAS Number:
Molecular Weight:
514.59
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥90% (HPLC)

form

powder

fluorescence

λex 526 nm; λem 640 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CCN(CC)c1ccc2C=C(\C=C\c3ccc(c[n+]3CCCCCC(O)=O)S([O-])(=O)=O)C(=O)Oc2c1

InChI

1S/C26H30N2O7S/c1-3-27(4-2)22-12-9-19-16-20(26(31)35-24(19)17-22)10-11-21-13-14-23(36(32,33)34)18-28(21)15-7-5-6-8-25(29)30/h9-14,16-18H,3-8,15H2,1-2H3,(H-,29,30,32,33,34)

InChI key

XWKHECGJHWMWTB-UHFFFAOYSA-N

General description

Mega dyes are specifically designed for multicolor techniques. Fluorescent red Mega 480 is characterized by an extremely large stoke′s shift between excitation and emission maxima. This dye is especially well suited for excitation by argon lasers or other short wavelength light sources and can be excited using the same light source or filter settings as for fluorescein and other widely used dyes.

Application

Fluorescent red Mega 480 and other Mega dyes are specifically designed for multicolor techniques. It is characterized by an extremely large stoke′s shift between excitation and emission maxima. This dye is especially well suited for excitation by argon lasers or other short wavelength excitation light. This also means, that it can be excited with the same light source or filter arrangement than fluorescein and other widely used dyes as well as other Mega dyes. Due to the different emission wavelength several different labels can be detected independently in the same experiment. Due to the small size of the Mega-labels and the direct conversion of excitation light into emission without using a FRET mechanism we foresee a variety of possible applications including DNA sequencing and FISH microscopy.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service