Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

40723

Sigma-Aldrich

1,2-Dieicosapentaenoyl-sn-glycero-3-phosphocholine

≥99.0% (TLC)

Synonym(s):

PC, 1,2-Di-(5Z,8Z,11Z,14Z,17Z-eicosapentaenoyl)-sn-glycero-3-phosphocholine, EPAPC, PC(20:5(5Z,8Z,11Z,14Z,17Z)/20:5(5Z,8Z,11Z,14Z,17Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H76NO8P
CAS Number:
Molecular Weight:
826.09
UNSPSC Code:
12352211
PubChem Substance ID:

biological source

synthetic

Assay

≥99.0% (TLC)

form

paste

functional group

phospholipid

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

O=C(CCC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)OC[C@H](COP(OCC[N+](C)(C)C)([O-])=O)OC(CCC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC)=O

InChI

1S/C48H76NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-47(50)54-44-46(45-56-58(52,53)55-43-42-49(3,4)5)57-48(51)41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h8-11,14-17,20-23,26-29,32-35,46H,6-7,12-13,18-19,24-25,30-31,36-45H2,1-5H3/b10-8-,11-9-,16-14-,17-15-,22-20-,23-21-,28-26-,29-27-,34-32-,35-33-/t46-/m1/s1

InChI key

WAIHGVAZVKEQGR-YWTDJUEZSA-N

General description

Though unusual in higher animals, di-polyunsaturated-fatty-acid phosphatidylcholine (diPUFA-PtdCho) is produced at high abundance by the roundworm Caenorhabditis elegans, especially when grown at below normal temperature. C. elegans acyltransferase is unusual in exhibiting no preference for saturated over unsaturated fatty acids.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Manca et al.
Journal of pharmaceutical and biomedical analysis, 176, 112764-112764 (2019-08-12)
An accurate and specific gas chromatography-mass spectrometry (GC-MS) method was optimized to quantify specific polyunsaturated fatty acids (PUFAs) in plasma and in erythrocyte membranes for clinical purposes. The developed and fully-validated method showed optimal linearity in addition to adequate results
A Rosa et al.
Food & function, 11(6), 5179-5191 (2020-05-22)
Eupatilin is a dietary flavonoid isolated from the alpine wormwoods, used for the genepy liqueur production. This flavone protects cells and tissues against oxidative stress and targets cancer cells, inducing cytotoxicity, cell circle arrest, apoptosis and mitochondrial dysfunction. This study
A Rosa et al.
Food & function, 10(3), 1629-1642 (2019-03-06)
The dietary sesquiterpene dienone zerumbone (ZER) selectively targets cancer cells, inducing mitochondrial dysfunction and apoptosis, and protects non-cancerous cells towards oxidative stress and insult. This study examines the in vitro effects of ZER on lipid peroxidation in biological systems (cholesterol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service