Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

406RA-05A

Sigma-Aldrich

Human Osteoblasts-Rheumatoid Arthritis: HOb-RA, adult

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,590.00

$1,590.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
$1,590.00

About This Item

UNSPSC Code:
41106514

$1,590.00


Please contact Customer Service for Availability

biological source

human bone (rheumatoid arthritic)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 46

morphology

osteoblast

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

arthritis

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Human Osteoblasts-Rheumatoid Arthritis, HOb-RA, are isolated from bone of patients with rheumatoid arthritis (RA) and provide an excellent model system to study the patophisiology of that disease in vitro, including matrix mineralization, effects of inflammatory mediators and bone morphogenic proteins, physiological control of bone remodeling and regulation of bone metabolism.Characterization: Positive for bone mineralization.

Cell Line Origin

Bone

Application

model system to study the pathophisiology of rheumatoid arthritis, matrix mineralization, effects of inflammatory mediators, bone morphogenic proteins, bone remodeling and metabolism

Components

Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 10 doublings

Subculture Routine

Please refer to the HOb-RA Culture Protocol.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service