Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39791

SAFC

1-(2-Isothiocyanatoethyl)-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium bromide

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16BrN3O2S
CAS Number:
Molecular Weight:
418.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

Assay

≥90% (HPLC)

fluorescence

λex 401 nm; λem 579 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[Br-].COc1ccc(cc1)-c2cnc(o2)-c3cc[n+](CCN=C=S)cc3

InChI

1S/C18H16N3O2S.BrH/c1-22-16-4-2-14(3-5-16)17-12-20-18(23-17)15-6-9-21(10-7-15)11-8-19-13-24;/h2-7,9-10,12H,8,11H2,1H3;1H/q+1;/p-1

InChI key

SKICOJPEBUOGMX-UHFFFAOYSA-M

General description

Class of highly photostable, water soluble fluorescent labels with large Stoke′s shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient (Ε∼ >20,000 in EtOH).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Litak, P.T.;Kauffman, J.M.
Journal of Heterocyclic Chemistry, 31, 457-457 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service