Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

39753

Supelco

N-(4-Isothiocyanatobenzyl)-4-[5-(4-methoxyphenyl)-2-oxazolyl]pyridinium bromide

suitable for fluorescence, BioReagent, ≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H18BrN3O2S
CAS Number:
Molecular Weight:
480.38
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Quality Level

Assay

≥98.0% (HPLC)

fluorescence

λem 569 nm

suitability

suitable for fluorescence

storage temp.

−70°C

SMILES string

[Br-].COc1ccc(cc1)-c2cnc(o2)-c3cc[n+](Cc4ccc(cc4)N=C=S)cc3

InChI

1S/C23H18N3O2S.BrH/c1-27-21-8-4-18(5-9-21)22-14-24-23(28-22)19-10-12-26(13-11-19)15-17-2-6-20(7-3-17)25-16-29;/h2-14H,15H2,1H3;1H/q+1;/p-1

InChI key

DMAVLRVUKGJFEU-UHFFFAOYSA-M

General description

Class of highly photostable, water soluble fluorescent labels with large Stoke′s shift, high QY in aqueous media and pH tolerance. Relatively low extinction coefficient.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Litak, P.T.;Kauffman, J.M.
Journal of Heterocyclic Chemistry, 31, 457-457 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service