Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

386R-1

Sigma-Aldrich

MyoD1 (EP212) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$267.00
1 ML
$267.00
0.1 ML
$339.00
0.1 ML
$339.00
0.5 ML
$850.00
0.5 ML
$850.00
7 ML
$898.00
7 ML
$898.00
1 ML
$1,270.00
1 ML
$1,270.00

$267.00


Available to ship onMarch 28, 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
$267.00
1 ML
$267.00
0.1 ML
$339.00
0.1 ML
$339.00
0.5 ML
$850.00
0.5 ML
$850.00
7 ML
$898.00
7 ML
$898.00
1 ML
$1,270.00
1 ML
$1,270.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$267.00


Available to ship onMarch 28, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP212, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (386R-14)
vial of 0.5 mL concentrate (386R-15)
bottle of 1.0 mL predilute (386R-17)
vial of 1.0 mL concentrate (386R-16)
bottle of 7.0 mL predilute (386R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG

control

rhabdomyosarcoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

human ... MYOD1(4654)

General description

MyoD1, one of the MyoD family of myogenic helix-loop-helix transcription factors, combinedwith myogenin, plays a role in coordinating the differentiation of skeletal muscle. MyoD1 isexpressed in myoblasts before differentiation. Anti-MyoD1 immunostaining identifies cellscommitted to myogenesis in their earliest phase. In conjunction with other markers, antiMyoD1 may be used for the identification of rhabdomyosarcoma.

Quality


IVD

IVD

IVD

RUO

Linkage

MyoD1 Positive Control Slides, Product No. 386S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raffaella A Morotti et al.
The American journal of surgical pathology, 30(8), 962-968 (2006-07-25)
Immunohistochemistry remains the current ancillary method of choice in the pathologic evaluation of small blue round-cell tumors. In at least 20% of cases of rhabdomyosarcoma (RMS), it is considered an essential factor in the final and/or differential diagnosis of the
N J Sebire et al.
Journal of clinical pathology, 56(6), 412-416 (2003-06-05)
The diagnosis of paediatric solid tumours is often based on small tissue needle biopsies in which many different entities demonstrate a "small round cell tumour" phenotype and in which there may be insufficient tissue to allow the interpretation of diagnostic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service