Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

38366

Sigma-Aldrich

Lipase, immobilized in Sol-Gel-AK, from Pseudomonas fluorescens

≥30 U/g

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
EC Number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Pseudomonas fluorescens

form

powder

specific activity

≥30 U/g

storage temp.

2-8°C

Unit Definition

1 U corresponds to the amount of enzyme which liberates 1 μmol oleic acid per minute at pH 8.0 and 40°C (triolein, Cat. No. 62314 as substrate)

Other Notes

Immobilized lipase with excellent characteristics for enzyme-catalyzed reactions

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.T. Reetz et al.
Angewandte Chemie (International Edition in English), 107, 373-373 (1995)
Takanori Matsui et al.
Biochemical and biophysical research communications, 443(3), 847-851 (2013-12-18)
Pigment epithelium-derived factor (PEDF) has anti-inflammatory and anti-thrombogenic properties both in cell culture and animal models. Although adipose triglyceride lipase (ATGL) and laminin receptor (LR) are two putative receptors for PEDF, which receptor mainly mediates the beneficial effects of PEDF
Sang Hoon Lee et al.
Natural product communications, 8(10), 1439-1441 (2013-12-21)
Obesity, which is characterized by excessive fat accumulation, occurs by fat absorption by lipase and sequential fat accumulation in adipocytes through adipocyte differentiation. Thus, inhibition of pancreatic lipase activity and adipocyte differentiation would be crucial for the prevention and treatment
Amit G Singal et al.
The American journal of gastroenterology, 109(3), 325-334 (2014-01-22)
The PNPLA3 rs738409 single-nucleotide polymorphism is known to promote nonalcoholic steatohepatitis (NASH), but its association with fibrosis severity and hepatocellular carcinoma (HCC) risk is less well-defined. The objectives of this study were to determine the association between PNPLA3 and liver
Ken-ichi Hirano et al.
Biochemical and biophysical research communications, 443(2), 574-579 (2013-12-18)
Adipose triglyceride lipase (ATGL, also known as PNPLA2) is an essential molecule for hydrolysis of intracellular triglyceride (TG). Genetic ATGL deficiency is a rare multi-systemic neutral lipid storage disease. Information regarding its clinical profile and pathophysiology, particularly for cardiac involvement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service