Skip to Content
MilliporeSigma
All Photos(1)

Documents

349M-9

Sigma-Aldrich

ZAP-70 (2F3.2) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

2F3.2, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (349M-94)
vial of 0.5 mL concentrate (349M-95)
bottle of 1.0 mL predilute (349M-97)
vial of 1.0 mL concentrate (349M-96)
bottle of 7.0 mL predilute (349M-98)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG2a

control

chronic lymphocytic leukemia

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... ZAP70(7535)

General description

Zeta-associated protein-70 (ZAP-70), a member of the Syk family of tyrosine kinases, has an important role in T-cell receptor signaling, natural killer cell activation, and early B-cell development. ZAP-70 protein is not expressed in most normal mature B-cells, but is expressed in various B- and T-cell lymphomas. Attempts to implement ZAP-70 analysis in clinical flow cytometry laboratories have been fraught with difficulty; this approach is technically challenging and results often have been unreliable. Detection of ZAP-70 protein by immunohistochemistry using tissue sections of chronic lymphocytic leukemia/small lymphocyctic lymphoma (CLL/SLL) is an easy and inexpensive technique. The clinical course of CLL/SLL is extremely variable: in some patients it is aggressive and requires chemotherapy within a relatively short time of diagnosis; in others it is indolent and asymptomatic and may not require treatment for many years. It has recently been demonstrated that the mutational status of the immunoglobulin heavy-chain variable-region (IgVH) genes identifies 2 distinct groups of CLL/SLL: the patients with unmutated IgVH have an unfavorable clinical course, whereas those with mutated IgVH experience a favorable course. The reasons for the differences in the clinical course of CLL patients with mutated and unmutated IgVH genes are not clear. ZAP-70 expression is the best discriminator of mutated and unmutated IgVH genes, ZAP-70 positive (>20% tumor cells) being Ig-unmutated and ZAP-70 negative being Ig-mutated. Wiestner et al. demonstrated that ZAP-70 expression levels assessed by means of Western blotting, real-time RT-PCR and immunohistochemistry are closely correlated with IgVH mutational status.

Quality


IVD

IVD

IVD

NA

Linkage

ZAP-70 Positive Control Slides, Product No. 349S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liguang Chen et al.
Blood, 100(13), 4609-4614 (2002-10-24)
We examined isolated leukemia B cells of patients with chronic lymphocytic leukemia (CLL) for expression of zeta-associated protein 70 (ZAP-70). CLL B cells that have nonmutated immunoglobulin variable region genes (V genes) expressed levels of ZAP-70 protein that were comparable
Marta Crespo et al.
The New England journal of medicine, 348(18), 1764-1775 (2003-05-02)
The mutational status of immunoglobulin heavy-chain variable-region (IgVH) genes in the leukemic cells of chronic lymphocytic leukemia (CLL) is an important prognostic factor in the disease. We investigated whether the expression of ZAP-70 by CLL cells correlated with the IgVH
Adrian Wiestner et al.
Blood, 101(12), 4944-4951 (2003-02-22)
The presence or absence of somatic mutations in the expressed immunoglobulin heavy chain variable regions (IgVH) of chronic lymphocytic leukemia (CLL) cells provides prognostic information. Patients whose leukemic cells express unmutated IgVH regions (Ig-unmutated CLL) often have progressive disease, whereas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service