Skip to Content
MilliporeSigma
All Photos(1)

Documents

3446-005-01

Sigma-Aldrich

Cultrex 3-D Culture Matrix Laminin I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

mouse (Engelbreth-Holm-Swarm (EHS) mouse tumor)

sterility

sterile

Assay

≥90% (SDS-PAGE)

concentration

6 mg/mL

technique(s)

cell culture | mammalian: suitable

solubility

water: miscible

shipped in

dry ice

storage temp.

−70°C

General description

3-D Culture Matrix Mouse Laminin I may be used as a gel on which to grow cells, or as a media additive alone or in concert with other basement membrane components, to study cellular growth and differentiation in three dimensions in vitro. To offer the most standardized Laminin I for use in 3-D cultures, a special process is employed to provide material at a standard concentration of 6 mg/ml (by absorbance and extinction coefficient).

Physical form

Solution in Dulbecco′s Modified Eagle′s medium containing 10 μg/ml gentamycin sulfate and no phenol red.

Other Notes

Functional Assay:
3-D Culture: Laminin I promotes differentiation of a human epithelial cell line derived from mammary gland (MCF-10A) into acinar structures.
Cell Attachment: Tested for the ability to promote cell attachment and spreading of MG63 human osteosarcoma cells.
Sterility Testing:
No bacterial or fungal growth detected after incubation at 37°C for 14 days following USP XXIV Chapter 71 sterility test.No mycoplasma contamination detected by PCR.Endotoxin concentrations = 20 EU/ml by LAL assay.
Storage Conditions and Product Stability:
Product is stable for a minimum of 3 months from date of shipment when stored at -80 °C in a manual defrost freezer. For optimal stability, store at -80 °C in aliquots. Avoid repeated freeze/thaw cycles.

Legal Information

3-D Culture Matrix is a trademark of Trevigen, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Rüdiger et al.
Cell reports, 32(6), 108015-108015 (2020-08-14)
Endothelial tube formation on a reconstituted basement membrane (Matrigel) is a well-established in vitro model for studying the processes of angiogenesis and vasculogenesis. However, to date, the organizing principles that underlie the morphogenesis of this network and that shape the initial
Ruilin Tian et al.
Neuron, 104(2), 239-255 (2019-08-20)
CRISPR/Cas9-based functional genomics have transformed our ability to elucidate mammalian cell biology. However, most previous CRISPR-based screens were conducted in cancer cell lines rather than healthy, differentiated cells. Here, we describe a CRISPR interference (CRISPRi)-based platform for genetic screens in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service