Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31393

Sigma-Aldrich

Dextran blue from Leuconostoc spp.

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H10O5]n
Molecular Weight:
162.14 (as monomer)
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
To inquire about this product 31393, please contact your local MilliporeSigma office or dealer. Contact Technical Service

biological source

bacterial (Leuconostoc spp.)

form

powder or crystals

mol wt

Mr ~2000000

storage temp.

room temp

Application

Dextran blue may be used as a macromolecular model protein in the preparation and characterization of biostructures such as microspheres. Dextran blue binds lactate dehydrogenase isoenzyme-5 (LDH-5) and may be used in assay development. Dextran blue is used in various tissue diffusion assays.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Preparation and characterization of protein loaded microspheres based on a hydroxylated aliphatic polyester, poly(lactic-co-hydroxymethyl glycolic acid).
Ghassemi AH, van Steenbergen MJ, et al.
J. Controlled Release, 138, 57-63 (2009)
M U Javed et al.
Experimental & molecular medicine, 33(2), 103-105 (2001-07-20)
Albumin is the most abundant protein in human serum. A dye-binding method is commonly used in clinical laboratories for its estimation using different types of dyes. However, all these dye methods were interfered by a variety of compounds. Here we
Clarise Rivera Starr et al.
Infection and immunity, 74(1), 40-48 (2005-12-22)
Group A streptococcus (GAS) depends on a hyaluronic acid (HA) capsule to evade phagocytosis and to interact with epithelial cells. Paradoxically, GAS also produces hyaluronidase (Hyl), an enzyme that cleaves HA. A common assumption is that Hyl digests structurally identical

Articles

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service