Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

312010

Sigma-Aldrich

Bromobenzene-UL-14C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
14C6H5Br
CAS Number:
Molecular Weight:
157.01
MDL number:
UNSPSC Code:
12352200

extent of labeling

1-15 mCimmol

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Pagliero et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(49), 19510-19515 (2011-11-22)
We use optical Faraday rotation (OFR) to probe nuclear spins in real time at high-magnetic field in a range of diamagnetic sample fluids. Comparison of OFR-detected NMR spectra reveals a correlation between the relative signal amplitude and the fluid Verdet
Heibatullah Kalantari et al.
Human & experimental toxicology, 30(10), 1710-1715 (2011-01-21)
The efficacy of a crude hydro-alcoholic extract of Cassia fistula (golden shower tree) fruit to protect the kidney against bromobenzene-induced toxicity was studied. Negative control mice received normal saline; positive control mice were given 460 mg/kg of bromobenzene; Cassia fistula
Jonell N Smith et al.
The Analyst, 136(1), 120-127 (2010-10-27)
Gaseous ammonia, a common toxic industrial compound, is not detected readily in ion trap mass spectrometers because its molecular ion falls below the low-mass cutoff (~m/z 40) normally used when examining organic compounds. Instead, reactions of ammonia with halobenzene radical
Maria J Nueda et al.
Biostatistics (Oxford, England), 13(3), 553-566 (2011-11-17)
Transcriptomic profiling experiments that aim to the identification of responsive genes in specific biological conditions are commonly set up under defined experimental designs that try to assess the effects of factors and their interactions on gene expression. Data from these
Sergey Vshyvenko et al.
Bioorganic & medicinal chemistry letters, 21(16), 4750-4752 (2011-07-16)
The synthesis of two C-1 analogues of pancratistatin has been accomplished in 17 steps from bromobenzene. The key steps involved the enzymatic dihydroxylation, regioselective opening of epoxyaziridine 9 with the alane derived from 8, a solid-state silica-gel-catalyzed intramolecular opening of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service