Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30693

Sigma-Aldrich

Chromeo P503

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$830.00

$830.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$830.00

About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

$830.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

fluorescence

λex 503 nm; λem 610 nm±20 nm in 0.1 M bicarbonate buffer pH 8.3

storage temp.

−20°C

General description

Chromeo P503 labels proteins and peptides by exhibiting a color change from blue to red upon binding to primary amines. Chromeo P503 displays a weak fluorescence with a quantum yield <1% in solution. After conjugation to a primary amine group, the label undergoes a shortwave spectral shift of >100 nm and the quantum yield rises to 50%. This property allows a distinct detection of primary amines, proteins, and other biomolecules.

Application

Chromeo P503 is used as a fluorogenic reagent to label primary amine groups within molecules such as proteins. Chromeo P503 labeled proteins may be studied in a variety of gel electrophoresis and chromatographic applications.

Caution

To ensure stability, the lyophilized dye should be stored at 4°C in the dark. This product is guaranteed for 6 months from the date of arrival.

Legal Information

Chromeo is a trademark of Active Motif Chromeon GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jane A Dickerson et al.
Electrophoresis, 31(15), 2650-2654 (2010-07-07)
CIEF and CZE are coupled with LIF detection to create an ultrasensitive 2-D separation method for proteins. In this method, two capillaries are joined through a buffer-filled interface. Separate power supplies control the potential at the injection end of the
Emily H Turner et al.
Journal of chromatography. A, 1194(2), 253-256 (2008-05-17)
The spectroscopic and electrophoretic properties of proteins labeled with Chromeo P503 were investigated. Its photobleaching characteristics were determined by continually infusing Chromeo P503-labeled alpha-lactalbumin into a sheath-flow cuvette and monitored fluorescence as a function of laser power. The labeled protein
Stephanie de Jong et al.
Analytical chemistry, 83(16), 6330-6335 (2011-07-07)
Methods of kinetic capillary electrophoresis (KCE) facilitate highly efficient selection of DNA aptamers for protein targets. The inability to detect native proteins at low concentrations in capillary electrophoresis creates, however, a significant obstacle for many important protein targets. Here we
Lauren M Ramsay et al.
Electrophoresis, 30(2), 297-302 (2009-02-11)
We have coupled CIEF with an LIF detector that is based on a post-column sheath flow cuvette. We employed Chromeo P503 as a fluorogenic reagent to label proteins before analysis. This reagent reacts with the epsilon-amine of lysine residues, preserving

Questions

  1. What is the solubility of this product in Methanol?

    1 answer
    1. This compound is tested for solubility at 10 mg/mL in Dimethylformamide. Methanol and acetonitrile are also listed as solvents, but a rate has not been established. Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/425/192/30693dat.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service