Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30564

Sigma-Aldrich

Trehalose 6-hexadecanoate

≥95.0% (GC)

Synonym(s):

α,α-Trehalose 6-palmitate, α-D-Glucopyranosyl-α-D-glucopyranoside 6-hexadecanoate, 6-O-Palmitoyl-α,α-trehalose, Trehalose 6-palmitate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H52O12
CAS Number:
Molecular Weight:
580.71
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95.0% (GC)

form

powder

mol wt

580.71 g/mol

CMC

0.0061 mM

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@H]1O[C@H]2O[C@H](COC(CCCCCCCCCCCCCCC)=O)[C@@H](O)[C@H](O)[C@H]2O

InChI

1S/C28H52O12/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-20(30)37-17-19-22(32)24(34)26(36)28(39-19)40-27-25(35)23(33)21(31)18(16-29)38-27/h18-19,21-29,31-36H,2-17H2,1H3/t18?,19?,21-,22-,23?,24?,25+,26+,27-,28-/m1/s1

InChI key

VNHFMAKRYDWMAF-DHFWNHDGSA-N

Application

Trehalose is commonly used as a stabilizer and protective material for biomaterial compounds. Because of these characteristics, Trehalose is introduced as a hydrophilic part. Due to these properties of Trehalose, protein degeneration is expected to be prevented with Trehalose detergents. Therefore, Trehalose detergents may keep the function of the protein and be utilized in the field of proteomics research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nawal K Paul et al.
The Journal of organic chemistry, 78(2), 363-369 (2012-12-12)
It was shown that reaction of trehalose with 1 equiv of a fatty acid in pyridine promoted by 1 equiv of the uronium-based coupling agent 2-(1H-benzotriazole-1-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate (TBTU) at room temperature gives a good yield of the primary ester accompanied
A K Datta et al.
Carbohydrate research, 218, 95-109 (1991-09-30)
A simplified synthesis of 6-mono- and 6,6'-di-corynomycolate esters of alpha,alpha-trehalose, and related compounds, was achieved by coupling the (hydroxyl-protected) acids to the partially trimethylsilylated sugar in the presence of dicyclohexylcarbodiimide and 4-dimethylaminopyridine. As acid reactants, (2-RS,3-RS)-3-hydroxy-2-tetradecyloctadecanoic acid (DL-corynomycolic acid) and
L Schiefelbein et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 76(3), 342-350 (2010-09-08)
Nonionic polyethylene glycol-derived surfactants are today's choice as surfactants in protein formulations. Different groups discovered that although surface-induced stresses are reduced by these excipients, the long-term stability of different proteins decreased due to polyethylene glycol-related induction of oxidation processes under
Chemical and biochemical studies on carbohydrate esters. V. Anti Ehrlich ascites tumor effect and chromatographic behaviors of fatty acyl monoesters of sucrose and trehalose.
Y Nishikawa et al.
Chemical & pharmaceutical bulletin, 25(7), 1717-1724 (1977-07-01)
Surface activities of monoacyl trehaloses in aqueous solution.
Chen, J., et al.
Food Sci. Technol., 40, 412-417 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service