Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

29297

Sigma-Aldrich

N-[2-(2-Aminoethylamino)ethyl]rhodamine 6G-amide bis(trifluoroacetate)

BioReagent, suitable for fluorescence, ≥90% (HPCE)

Synonym(s):

Rhodamine 6G-N-[2-(2-aminoethylamino)ethyl]amide bis(trifluoroacetate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H37N5O2·C4H2F6O4
Molecular Weight:
727.69
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.25

product line

BioReagent

Quality Level

Assay

≥90% (HPCE)

fluorescence

λex 528 nm; λem 553 nm in 0.1 M phosphate pH 3.0

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.CCNc1cc2OC3=CC(=N\CC)\C(C)=CC3=C(c2cc1C)c4ccccc4C(=O)NCCNCCN

InChI

1S/C30H37N5O2.2C2HF3O2/c1-5-33-25-17-27-23(15-19(25)3)29(24-16-20(4)26(34-6-2)18-28(24)37-27)21-9-7-8-10-22(21)30(36)35-14-13-32-12-11-31;2*3-2(4,5)1(6)7/h7-10,15-18,32-33H,5-6,11-14,31H2,1-4H3,(H,35,36);2*(H,6,7)/b34-26-;;

InChI key

YKXRITLMCRSWKA-IQKNAWRNSA-N

Application

N-[2-(2-Aminoethylamino)ethyl]rhodamine 6G-amide bis(trifluoroacetate) is a rhodamine 6G-amide derivative useful as a bioreagent in fluorescence assays.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Yuan et al.
Organic & biomolecular chemistry, 9(6), 1723-1726 (2011-01-25)
A novel strategy to systematically tune the pK(a) values of rhodamines is described. This strategy was applied to rationally develop compound 1e with a pK(a) of 6.5, the highest among rhodamine amide derivatives, and it could be employed to detect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service