Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

262R-1

Sigma-Aldrich

Granzyme B (EP230) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$225.00
0.1 ML
$277.00
7 ML
$666.00
1 ML
$1,160.00

$225.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$225.00
0.1 ML
$277.00
7 ML
$666.00
1 ML
$1,160.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$225.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP230, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (262R-14)
vial of 0.5 mL concentrate (262R-15)
bottle of 1.0 mL predilute (262R-17)
vial of 1.0 mL concentrate (262R-16)
bottle of 7.0 mL predilute (262R-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

control

anaplastic large cell lymphoma, spleen

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... GZMB(3002)

General description

Granzymes are serine proteases which are stored in specialized lytic granules of cytotoxic T-lymphocytes and in natural killer cells.1 Anti-granzyme B has been useful in in the identification of natural killer/T-cell lymphoma, as well as anaplastic large cell lymphoma.2-4

Quality


IVD

IVD

IVD

RUO

Linkage

Granzyme B Positive Control Slides, Product No. 262S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoko Kato et al.
The American Journal of dermatopathology, 25(2), 142-147 (2003-03-26)
A neutrophil-rich anaplastic large cell lymphoma (ALCL) presented in the skin of a 47-year-old Japanese woman. The solitary cutaneous lesion was an eroded, 10-mm, dome-shaped nodule involving the skin of her left upper arm. Histologically, it showed a proliferation of
J A Kummer et al.
Clinical and experimental immunology, 100(1), 164-172 (1995-04-01)
Cytoplasmic granules from activated natural killer (NK) and cytotoxic T lymphocytes (CTL) contain a pore-forming protein, perforin, and several homologous serine proteinases called granzymes. Expression of these proteins correlates with the cytolytic potential of cytotoxic lymphocytes. Using a panel of
J J Oudejans et al.
The American journal of pathology, 148(1), 233-240 (1996-01-01)
Reed-Sternberg (RS) and Hodgkin's (H) cells are considered to be the neoplastic cells in Hodgkin's disease. Although most data suggest a lymphoid origin, the nature of these cells still remains the subject of considerable controversy. Recently, monoclonal antibodies became available
Jie Liu et al.
The Journal of dermatology, 30(10), 735-741 (2003-12-20)
Nasal natural killer (NK)/T cell lymphoma is an Epstein-Barr virus (EBV) associated lymphoma that arises in the nasal area and aggressively invades surrounding tissues. Our patient was a 48-year-old male who had had nasal obstruction and nasal discharge for 2
Brett E Johnson et al.
Cell reports. Medicine, 3(2), 100525-100525 (2022-03-05)
Mechanisms of therapeutic resistance and vulnerability evolve in metastatic cancers as tumor cells and extrinsic microenvironmental influences change during treatment. To support the development of methods for identifying these mechanisms in individual people, here we present an omic and multidimensional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service