Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

226R-2

Sigma-Aldrich

BCL2 (SP66) Rabbit Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SP66, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (226R-24)
vial of 0.5 mL concentrate (226R-25)
bottle of 1.0 mL predilute (226R-27)
vial of 1.0 mL concentrate (226R-26)
bottle of 7.0 mL predilute (226R-28)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... BCL2(596)

General description

BCL2 is a protein associated with apoptosis regulation produced by the bcl-2 gene, located on chromosome 14q32. BCL2 is comprised of an alpha (239 amino acids) and beta chain. BCL2 (and thus BCL2 alpha chain) is found in mitochondrial and nuclear membranes and in the cytosol rather than the cell surface. In normal lymphoid tissue, BCL2 (and BCL2 alpha) antibody reacts with small B-lymphocytes in the mantle zone and many cells within the T-cell areas. Anti-BCL2 alpha has shown consistent negative reaction on reactive germinal centers and positive staining of neoplastic follicles in follicular lymphoma. This difference in staining pattern is not due to down regulation or decreased BCL2 mRNA, but largely to a post-translational mechanism with resultant decrease in protein levels. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymphoid lesions. Anti-BCL2 has been used as an indicator of minimal residual disease in the bone marrow of follicular lymphoma patients when staining is strong and uniform.

Linkage

BCL2 Positive Control Slides, Product No. 226S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manual of Diagnostic Antibodies for Immunohistology, 25-27 (2003)
K Cooper et al.
The Journal of pathology, 182(3), 307-310 (1997-07-01)
Recent studies have shown bcl-2 to be regulated by p53. Other studies have suggested an inverse relationship between p53 and bcl-2 protein expression in breast and colonic cancers and in a variety of subtypes of non-Hodgkin's lymphoma. This study investigates
R Chetty et al.
Journal of clinical pathology, 48(11), 1035-1038 (1995-11-01)
To establish the role of immunohistochemistry (using a limited panel of antibodies) in detecting minimal involvement by follicular lymphoma in routinely processed bone marrow trephine specimens, which show no obvious morphological (light microscopic) evidence of lymphoma; to determine whether bcl-2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service