Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

211R-1

Sigma-Aldrich

TSH (EP254) Rabbit Monoclonal Primary Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$202.00
0.1 ML
$245.00
7 ML
$494.00
0.5 ML
$692.00
1 ML
$1,110.00

$202.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$202.00
0.1 ML
$245.00
7 ML
$494.00
0.5 ML
$692.00
1 ML
$1,110.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$202.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP254, monoclonal

description

(For In Vitro Diagnostic Use in Select Regions (See Chart))

form

buffered aqueous solution

species reactivity

human

packaging

bottle of 1.0 mL predilute (211R-17)
bottle of 7.0 mL predilute (211R-18)
vial of 0.1 mL concentrate (211R-14)
vial of 0.5 mL concentrate (211R-15)
vial of 1.0 mL concentrate (211R-16)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

General description

Thyroid-stimulating hormone (also known as TSH or thyrotropin) is a peptide hormone synthesized and secreted by thyrotrope cells in the anterior pituitary gland which regulate the endocrine function of the thyroid gland. TSH is a glycoprotein and consists of two subunits, the alpha and the beta subunit, which are non-covalently bound to one another. The alpha subunit of TSH is also present in two other pituitary glycoprotein hormones: Follicle stimulating hormone and luteinizing hormone and, in primates, in the placental hormone chorionic gonadotropin. Each of these hormones also has a unique beta subunit, which provides receptor specificity. In other words, TSH is composed of alpha subunit bound to the TSH beta subunit, and TSH associates only with its own receptor. Free alpha and beta subunits have essentially no biological activity. Anti-TSH reacts with TSH-producing cells (thyrotrophs), and is a useful marker in classification of pituitary tumors and the differential identification of primary and metastatic tumors in the pituitary gland.1-5

Quality


IVD

IVD

IVD

RUO

Linkage

TSH Positive Control Slides, Product No. 211S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J N Clore et al.
The American journal of the medical sciences, 295(1), 3-5 (1988-01-01)
A 36-year-old woman with hyperthyroidism, elevated blood thyroid-stimulating hormone (TSH) and alpha-subunit levels, amenorrhea, hyperprolactinemia and no evidence of acromegaly, was found to have a pituitary adenoma containing TSH, alpha-subunit and growth hormone by immunohistochemistry. Preoperative testing revealed elevated TSH
N Sanno et al.
The Journal of clinical endocrinology and metabolism, 80(8), 2518-2522 (1995-08-01)
TSH-secreting pituitary adenomas are rare. The transcriptional expression (messenger ribonucleic acids: mRNAs) of TSH beta, GH, and PRL in five patients with TSH-secreting pituitary adenoma was studied by the in situ hybridization (ISH) method in order to elucidate their multiple
E Batanero et al.
Brain, behavior, and immunity, 6(3), 249-264 (1992-09-01)
We evaluated the presence of anterior pituitary hormones; follicle-stimulating hormone (FSH) and its beta-subunit (beta-FSH), luteinizing hormone (LH) and its beta-subunit (beta-LH), beta-subunit of thyroid-stimulating hormone (beta-TSH), adrenocorticotropic hormone (ACTH), growth hormone (GH), and prolactin (PRL); the placental hormone human
N Kuzuya et al.
The Journal of clinical endocrinology and metabolism, 71(5), 1103-1111 (1990-11-01)
Endocrine and immunohistochemical studies were performed in two cases of TSH-secreting pituitary adenomas. The patients had elevated serum TSH and alpha-subunit concentrations despite high serum thyroid hormone levels. In addition, one patient (no. 1) had elevated serum GH levels with
S La Rosa et al.
Virchows Archiv : an international journal of pathology, 437(3), 264-269 (2000-10-19)
Gonadotropin-releasing hormone (GnRH), which is a well-known regulator of gonadotroph function, has recently been considered to be a paracrine factor involved in the control of somatotroph, lactotroph, and corticotroph cells. GnRH action is initiated by binding to a specific cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service