Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

15396

Sigma-Aldrich

Boc-Sta(3R,4S)-OH

≥97.0% (sum of enantiomers, HPLC)

Synonym(s):

(3R,4S)-4-(Boc-amino)-3-hydroxy-6-methylheptanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H25NO5
CAS Number:
Molecular Weight:
275.34
Beilstein:
3613624
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥97.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D −26±2°, c = 0.5% in methanol

mp

137-139 °C

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](NC(=O)OC(C)(C)C)[C@H](O)CC(O)=O

InChI

1S/C13H25NO5/c1-8(2)6-9(10(15)7-11(16)17)14-12(18)19-13(3,4)5/h8-10,15H,6-7H2,1-5H3,(H,14,18)(H,16,17)/t9-,10+/m0/s1

InChI key

KJIQRHBVNGRPCO-VHSXEESVSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service