Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14782

Sigma-Aldrich

L-Fuculose 1-phosphate lithium salt

≥97% (TLC)

Synonym(s):

6-Deoxytagatose 1-phosphate lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13O8P · xLi+
CAS Number:
Molecular Weight:
244.14 (free acid basis)
UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥97% (TLC)

form

powder

storage temp.

−20°C

InChI

1S/C6H13O8P/c1-3(7)5(9)6(10)4(8)2-14-15(11,12)13/h3,5-7,9-10H,2H2,1H3,(H2,11,12,13)/p-2/t3-,5+,6-/m0/s1

InChI key

KNYGWWDTPGSEPD-LFRDXLMFSA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service