Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14772

Sigma-Aldrich

N,N′-Bis(2-dimethylaminoethyl)-N,N′-dimethyl-9,10-anthracenedimethanamine biszinc chloride complex

BioReagent, suitable for fluorescence, ≥90% (TLC)

Synonym(s):

9,10-Bis(TMEDA)anthracene biszinc chloride complex, 9,10-Bis[N-(2-dimethylaminoethyl)methylaminomethyl]anthracene biszinc chloride complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H38Cl4N4Zn2
CAS Number:
Molecular Weight:
679.20
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

product line

BioReagent

Assay

≥90% (TLC)

solubility

DMSO: soluble
acetonitrile: soluble

fluorescence

λex 335 nm; λem 427 nm in acetonitrile

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

Cl[Zn]Cl.Cl[Zn]Cl.CN(C)CCN(C)Cc1c2ccccc2c(CN(C)CCN(C)C)c3ccccc13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service