Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14700C

SAFC

EX-CELL® CD Hydrolysate Fusion

Animal-component free, liquid, sterile-filtered, suitable for cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$192.00
1000 ML
$334.00

$192.00


Please contact Customer Service for Availability


Select a Size

Change View
500 ML
$192.00
1000 ML
$334.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.71

$192.00


Please contact Customer Service for Availability

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

storage condition

protect from light

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

EX-CELL® CD Hydrolysate Fusion is a synthetic option to undefined hydrolysates used in cell culture processes, including both Batch and Fed-Batch modes. It is formulated to have minimal pH and osmolality impact upon addition. EX-CELL CD Hydrolysate Fusion is optimized for Chinese Hamster Ovary (CHO) cell lines but has also been shown to work well with other cell types (e.g. NS0 and Sp2/0). EX-CELL CD Hydrolysate Fusion is comprised only of animal-free, chemically defined components that were identified to be in hydrolysates (but synthetically produced) or are commonly found in cell culture media. In order to have broad applications, EX-CELL CD Hydrolysate Fusion was based on data generated from four hydrolysate types (soy, wheat gluten, yeast extract and meat).

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service