Skip to Content
MilliporeSigma
All Photos(1)

Documents

14479

Sigma-Aldrich

Phalloidin-Atto 490LS

suitable for fluorescence

Synonym(s):

Atto 490LS-Phalloidin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

Assay

≥75.0% (HPLC)

Quality Level

mol wt

Mw 1466 g/mol

manufacturer/tradename

ATTO-TEC GmbH

λ

in acetonitrile: water (1:1)

UV absorption

λ: 495-501 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 490LS is a new fluorescent label featuring an extraordinary large Stokes-Shift of 165 nm. Thus the emission spectrum is almost completely separated from its absorption spectrum, making the dye highly suitable for multiplexing experiments, in particular in combination with Atto 488 and Atto 514. Atto 490LS is very hydrophillic and shows excellent water solubility. The dye exhibits a relatively high fluorescence quantum yield, which is only slightly reduced after conjugation to biomolecules, e.g. proteins, even at high degrees of labeling (DOL). Atto 490LS is an anionic dye. After conjugation to a substrate the dye carries a net electrical charge of -1.
Phalloidin, a bicyclic heptapeptide, is a very strong binding reagent to actin. Fluorescent labeled phalloidin has become a useful tool to investigate the distribution of F-actin within the cytoskeleton of cells by fluorescence microscopy.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaomeng Zhang et al.
Oncogene, 39(30), 5267-5281 (2020-06-21)
Melanoma is a deadly form of skin cancer that accounts for a disproportionally large proportion of cancer-related deaths in younger people. Compared with most other skin cancers, a feature of melanoma is its high metastatic capacity, although the mechanisms that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service