Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13520

Sigma-Aldrich

S-Benzylisothiourea hydrochloride

purum, ≥98.0% (AT)

Synonym(s):

S-Benzylthiuronium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(SCH2C6H5)=NH · HCl
CAS Number:
Molecular Weight:
202.70
Beilstein:
3656719
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (AT)

form

solid

mp

172-177 °C

solubility

ethanol: soluble 1 g/10 mL, clear, colorless to faintly greenish-yellow

SMILES string

Cl.NC(=N)SCc1ccccc1

InChI

1S/C8H10N2S.ClH/c9-8(10)11-6-7-4-2-1-3-5-7;/h1-5H,6H2,(H3,9,10);1H

InChI key

WJAASTDRAAMYNK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

S-Benzylisothiourea reacts with ethyl-γ-bromoacetoacetate to yield 2-benzylthio-4-benzylthio-methyl-6-hydroxypyrimidine.

Application

S-Benzylisothiourea hydrochloride was used to study the preparation, physical properties and supramolecular crystal structure of the ionic compound bis-(S-Benzylthiuronium)chloranilate.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Reaction of Ethyl α-and γ-Bromoacetoacetates with S-Alkylisothioureas.
Dodson RM, et al.
Journal of the American Chemical Society, 72(7), 3281-3282 (1950)
Bis-(S-benzylthiuronium) chloranilate supramolecular crystal structure: Preparation and characterization.
Stergiou DV, et al.
Chemistry of Materials, 17(6), 1307-1312 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service