Skip to Content
MilliporeSigma
All Photos(1)

Documents

12326

Sigma-Aldrich

Fluorescent red 631 reactive

suitable for fluorescence, ≥60% (coupling to amines)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H48N3NaO11S2
Molecular Weight:
833.94
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

concentration

≥60% (coupling to amines)

fluorescence

λex 627 nm; λem 651 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

[Na+].CCN(CC)c1ccc2c(\C=C\C=C3/N(CCCS([O-])(=O)=O)c4ccc(cc4C3(C)CCCC(=O)ON5C(=O)CCC5=O)S([O-])(=O)=O)cc([o+]c2c1)C(C)(C)C

InChI

1S/C40H49N3O11S2.Na/c1-7-41(8-2)28-15-17-30-27(24-35(39(3,4)5)53-33(30)25-28)12-9-13-34-40(6,21-10-14-38(46)54-43-36(44)19-20-37(43)45)31-26-29(56(50,51)52)16-18-32(31)42(34)22-11-23-55(47,48)49;/h9,12-13,15-18,24-26H,7-8,10-11,14,19-23H2,1-6H3,(H-,47,48,49,50,51,52);/q;+1/p-1/b12-9+,34-13-;

InChI key

FLVFZXRJCNTEDK-ANZJOGBDSA-M

General description

Fluorescent red 631 reactive is a fluorescent label especially well suited for excitation by He-Ne-lasers (at 633 nm). Fluorescent red 631 reactive shows strong fluorescence (molar absorption 185 000 M-1cm-1)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service