Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12271

Sigma-Aldrich

2-Keto-3-deoxy-D-gluconic acid lithium salt

≥95% (TLC)

Synonym(s):

3-Deoxy-D-erythro-2-hexulosonic acid lithium salt, KDG, Lithium 2-keto-3-deoxy-D-gluconate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O6 · xLi+
CAS Number:
Molecular Weight:
178.14 (free acid basis)
UNSPSC Code:
12352204
NACRES:
NA.32

Assay

≥95% (TLC)

form

powder or crystals

storage temp.

−20°C

InChI

1S/C6H10O6/c7-2-5(10)3(8)1-4(9)6(11)12/h3,5,7-8,10H,1-2H2,(H,11,12)/t3-,5+/m0/s1

InChI key

WPAMZTWLKIDIOP-WVZVXSGGSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Metabolite in the pentose phosphate pathway, which generates NADPH and pentoses; KDG is an intermediate in the metabolism of glucose and gluconic acid in some pseudomonas strains, of uronic acids in E. coli, and also an important metabolite in the degradation of polygalacturonates in Erwinia chrysanthemi.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Caution

Hygroscopic.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Uronic acid metabolism in bacteria. I. Purification and properties of uronic acid isomerase in Escherichia coli.
G ASHWELL et al.
The Journal of biological chemistry, 235, 1559-1565 (1960-06-01)
A new pathway of uronic acid metabolism.
A ASHWELL et al.
Biochimica et biophysica acta, 30(1), 186-187 (1958-10-01)
G Condemine et al.
Journal of bacteriology, 165(3), 937-941 (1986-03-01)
Mutants of Erwinia chrysanthemi impaired in pectin degradation were isolated by chemical and Mu d(Ap lac) insertion mutagenesis. A mutation in the kduD gene coding for 2-keto-3-deoxygluconate oxidoreductase prevented the growth of the bacteria on polygalacturonate as the sole carbon
Carbohydrate metabolism by Pseudomonas fluorescens. III. Purification and properties of a 6-phosphogluconate dehydrase.
R KOVACHEVICH et al.
The Journal of biological chemistry, 213(2), 745-756 (1955-04-01)
Glucose and gluconic acid oxidation of Pseudomonas saccharophila.
N ENTNER et al.
The Journal of biological chemistry, 196(2), 853-862 (1952-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service