Skip to Content
MilliporeSigma

11615

Azo-Casein sodium salt

lyophilized

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352300
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

assay

98.0-102.0% NaOH basis (1 M, titration)

form

fibers

quality

lyophilized

technique(s)

electrophoresis: suitable

solubility

H2O: easily soluble

Application

Literature see 11610 Azo-Casein

Other Notes

Sales restrictions may apply


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library