Skip to Content
MilliporeSigma
All Photos(1)

Documents

1041

p-Nitrophenol Standard

Synonym(s):

4-Nitrophenol solution, p-Nitrophenol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11
EC Number:
MDL number:
UNSPSC Code:
51000000

IVD

for in vitro diagnostic use

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H

InChI key

BTJIUGUIPKRLHP-UHFFFAOYSA-N

Other Notes

10 μmol/mL (10 mmol/L). For use in calibration of Bessey-Lowry-Brock acid
and alkaline phosphatase methods.

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service