Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05420

Sigma-Aldrich

Albumin from human serum

lyophilized powder, ≥95.0% (agarose gel electrophoresis)

Synonym(s):

HSA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$1,080.00

$1,080.00


Please contact Customer Service for Availability


Select a Size

Change View
5 G
$1,080.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
eCl@ss:
42010201

$1,080.00


Please contact Customer Service for Availability

biological source

human

Assay

≥95.0% (agarose gel electrophoresis)

form

lyophilized powder

mol wt

monomer calculated mol wt 66478 Da

technique(s)

tissue culture: suitable

impurities

HIV I and HIVII, HCV and HBsAg, tested negative

pH

7.0±0.2

solubility

H2O: 50 mg/mL, clear to slightly hazy, yellow

UniProt accession no.

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

Gene Information

human ... ALB(213)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Albumins are soluble monomeric proteins found in the body fluids and tissues of animals and in some plant seeds. Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones. Serum albumins are also vital in regulating the colloidal osmotic pressures of blood.
Serum albumin functions as a carrier protein for steroids, fatty acids, and thyroid hormones, and is vital in regulating the colloidal osmotic pressures of blood. Albumin is also seen to bind to exogenous substances, particularly drugs (e.g., ibuprofen, warfarin), and strongly influence their pharmacokinetics. Oxidative stress leading to changes in the redox state of albumin has widely varied effects on its physiological function.

Other Notes

View more information on human serum albumin.
Review: Serum albumin[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Penelope Rice et al.
Journal of immunology (Baltimore, Md. : 1950), 174(6), 3676-3685 (2005-03-08)
We previously demonstrated that exposure to febrile-range hyperthermia (FRH) accelerates pathogen clearance and increases survival in murine experimental Klebsiella pneumoniae peritonitis. However, FRH accelerates lethal lung injury in a mouse model of pulmonary oxygen toxicity, suggesting that the lung may
R Tzoneva et al.
Journal of biomaterials science. Polymer edition, 19(7), 837-852 (2008-06-12)
Poly(ether imide) (PEI) membranes of which the surface was modified with carboxylic groups were tested in comparison to pure PEI and poly(ethylene terephtalate) (PET) for their ability to support attachment, growth and function of human umbilical vein endothelial cells (HUVEC)
Mousumi Bora et al.
Veterinary world, 9(9), 1028-1033 (2016-10-14)
The objective of this study was to screen the prevalence of contagious ecthyma (CE) among the goat population of Assam owing to its high prevalence rate. In this study, a total of 231 serum samples were collected from 12 districts
Marek Jankowski et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(35), 13074-13079 (2004-08-19)
Previous studies demonstrated the presence of oxytocin (OT) and oxytocin receptors (OTRs) in the heart. The present work provides results supporting a potential role of OT in cardiomyogenesis. Here, we show a maximal OT and OTR protein level in the
T. Peters, Jr.
Adv. Prot. Chem., 37, 161-161 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service