Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04677

Sigma-Aldrich

Rutinose

≥98.0% (HPLC)

Synonym(s):

6-O-(α-L-Rhamnopyranosyl)-D-glucose, 6-O-(6-Deoxy-α-L-mannopyranosyl)-D-glucose

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$502.00

$502.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$502.00

About This Item

Empirical Formula (Hill Notation):
C12H22O10
CAS Number:
Molecular Weight:
326.30
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

$502.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98.0% (HPLC)

form

solid

color

white to light beige

storage temp.

room temp

SMILES string

C[C@@H]1O[C@@H](OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O)[C@H](O)[C@H](O)[C@H]1O
C[C@@H]1O[C@@H](OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C12H22O10/c1-4-7(16)10(19)11(20)12(22-4)21-3-6(15)9(18)8(17)5(14)2-13/h2,4-12,14-20H,3H2,1H3/t4-,5-,6+,7-,8+,9+,10+,11+,12+/m0/s1

InChI key

CPYCUQCIDSHOHI-IFLAJBTPSA-N

Application

Rutinose, a rhamnose and glucose containing disaccharide, is used to study the composition and function of flavonoid glycosides (eg. Rutin). Rutinose is also studied as a possible sugar metabolite in plants.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yashwantsinh S Jadeja et al.
Magnetic resonance in chemistry : MRC, 55(6), 589-594 (2016-12-13)
Hesperidin is flavonoid molecule found in citrus fruits (Citrus reticulata), especially difficult to extract, classify and characterize. Present work is to study the unresolved relative configuration of Hesperidin through the dihedral angle, coupling constant and different NMR techniques. The Karplus
Hui-Yi Lin et al.
Journal of cellular physiology, 202(2), 579-590 (2004-08-19)
Flavonoids including the aglycones, hesperetin (HT; 5,7,3'-trihydroxy-4'-methoxy-flavanone), and naringenin (NE; 5,7,4'-trihydroxy flavanone) and glycones, hesperidin (HD; 5,7,3'-trihydroxy-4'-methoxy-flavanone 7-rhamnoglucoside) and naringin (NI; 5,7,4'-trihydroxy flavanone 7-rhamno glucoside), were used to examine the importance of rutinose at C7 on the inhibitory effects of
Stefan Jenkins et al.
Frontiers in microbiology, 8, 2618-2618 (2018-01-10)
Exometabolomics enables analysis of metabolite utilization of low molecular weight organic substances by soil bacteria. Environmentally-based defined media are needed to examine ecologically relevant patterns of substrate utilization. Here, we describe an approach for the construction of defined media using
Marisol Minig et al.
Biotechnology and applied biochemistry, 62(1), 94-100 (2014-04-05)
The diglycosidase, α-rhamnosyl-β-glucosidase, from Acremonium sp. DSM24697 was immobilized by adsorption and cross-linking onto polyaniline-iron (PI) particles. The immobilization yield and the immobilization efficiency were relatively high, 31.2% and 8.9%, respectively. However, the heterogeneous preparation showed lower stability in comparison
Pavel Bartl et al.
International journal of food sciences and nutrition, 66(5), 514-519 (2015-06-20)
The anthocyanin composition of blue (Triticum aestivum L., cv. Skorpion) and purple wheat (Triticum aethiopicum JAKUBZ cv. Abyssinskaja arrasajta cv. Abyssinskaja arrasajta), cultivated in the Czech Republic, and of the prepared whole blue and purple wheat bread was determined. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service