Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04554

Sigma-Aldrich

1-Acetylisatin

≥97.5% (GC)

Synonym(s):

1-Acetyl-1H-indole-2,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO3
CAS Number:
Molecular Weight:
189.17
Beilstein:
144609
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

Assay

≥97.5% (GC)

form

powder or crystals

SMILES string

CC(=O)N1C(=O)C(=O)c2ccccc12

InChI

1S/C10H7NO3/c1-6(12)11-8-5-3-2-4-7(8)9(13)10(11)14/h2-5H,1H3

InChI key

LPGDEHBASRKTDG-UHFFFAOYSA-N

Biochem/physiol Actions

Metabolite of tryptophan pathway; selective inhibition of carboxylesterases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Wang et al.
The Journal of organic chemistry, 70(10), 3850-3858 (2005-05-07)
[reaction: see text] Photoinduced reactions of 1-acetylisatin (IS) with diphenylacetylenes 1a-c, 1-(p-methoxyphenyl)propyne 2, and 1,4-diphenyl-1,3-butadiyne 3 gave beta,beta-disubstituted 3-alkylidene oxindoles 6-12 respectively via [2+2] cycloaddition of 3IS* with the alkyne and subsequent oxetene ring opening. Photoreactions of IS with phenylacetylenes
J Xue et al.
Organic letters, 2(17), 2583-2586 (2000-09-16)
[reaction: see text]Photoinduced reactions of 1-acetylisatin 1 with diphenylacetylenes 2a-2c afforded the corresponding 3-methylene-2-indolones 4a-4e and 5a-5e in 80-90% yields via a spirooxetene intermediate. Similar irradiation of 1 with phenylacetylene, on the other hand, resulted in efficient formation of two
Janice L Hyatt et al.
Journal of medicinal chemistry, 50(8), 1876-1885 (2007-03-24)
Carboxylesterases (CE) are ubiquitous enzymes thought to be responsible for the metabolism and detoxification of xenobiotics. Numerous clinically used drugs including Demerol, lidocaine, capecitabine, and CPT-11 are hydrolyzed by these enzymes. Hence, the identification and application of selective CE inhibitors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service