Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03567

Sigma-Aldrich

(S)-2-Piperazinecarboxylic acid dihydrochloride

≥98.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O2 · 2HCl
CAS Number:
Molecular Weight:
203.07
Beilstein:
7312035
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Assay

≥98.0% (T)

form

powder or crystals

optical activity

[α]20/D −5±1°, c = 1% in H2O

color

white to faint beige

application(s)

peptide synthesis

SMILES string

Cl[H].Cl[H].OC(=O)[C@@H]1CNCCN1

InChI

1S/C5H10N2O2.2ClH/c8-5(9)4-3-6-1-2-7-4;;/h4,6-7H,1-3H2,(H,8,9);2*1H/t4-;;/m0../s1

InChI key

WNSDZBQLMGKPQS-FHNDMYTFSA-N

Application

Preparation of (S)-piperazine-2-carboxylic acid, (R)-piperazine-2-carboxylic acid, and (S)-piperidine-2-carboxylic acid by kinetic resolution of the corresponding racemic carboxamides with stereoselective amidases in whole bacterial cells[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service