Skip to Content
MilliporeSigma
All Photos(1)

Documents

01705

Sigma-Aldrich

Acrylamide:N,N′-Methylenebisacrylamide 19:1

BioXtra, suitable for electrophoresis

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioXtra

technique(s)

electrophoresis: suitable

impurities

≤0.05% free acid

conductivity

<20 μS/cm at 20 °C (50 mg/ml water)

solubility

H2O: 50 mg/mL at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤100 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

suitability

in accordance for electrophoresis test

storage temp.

2-8°C

SMILES string

NC(=O)C=C.C=CC(=O)NCNC(=O)C=C

InChI

1S/C7H10N2O2.C3H5NO/c1-3-6(10)8-5-9-7(11)4-2;1-2-3(4)5/h3-4H,1-2,5H2,(H,8,10)(H,9,11);2H,1H2,(H2,4,5)

InChI key

OCQZXMCGTAWGEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Premixed acrylamide/methylenebisacrylamide (95:5) for the preparation of acrylamide gel and its use in gel electrophoresis

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An experimental study about hydrogels for the fabrication of optical fiber humidity sensors.
<BIG>Francisco J. Arregui</BIG>
Sensors and Actuators B, Chemical, 96, 165-172 (2003)
Preparation and properties of acrylamide gel for use in electrophoresis.
S RAYMOND et al.
Analytical biochemistry, 1, 391-396 (1960-12-10)
M Nordin-Andersson et al.
Cell biology and toxicology, 19(1), 43-51 (2003-03-29)
Basal cytotoxicity, morphological changes and alterations in cell physiological and neurochemical functions were studied in differentiated human neuroblastoma (SH-SY5Y) cells during exposure to acrylamide and during a subsequent recovery period after cessation of exposure. Acrylamide induced a 20% reduction in
Roongnapa Suedee et al.
Journal of chromatography. A, 1114(2), 239-249 (2006-03-15)
A temperature sensitive dopamine-imprinted polymer was prepared in 80% aqueous methanol solution by free-radical cross-linking co-polymerisation of methacrylic acid and acrylamide at 60 degrees C in the presence of N,N-methylene-bis-acrylamide as the cross-linker and dopamine hydrochloride as template molecule. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service