Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

01702

Sigma-Aldrich

Acrylamide:N,N′-Methylenebisacrylamide 29:1

BioXtra, for molecular biology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319
NACRES:
NB.22

grade

Molecular Biology
for molecular biology

product line

BioXtra

technique(s)

electrophoresis: suitable

impurities

DNases, none detected
RNases, none detected
phosphatases, none detected
proteases, none detected

conductivity

<20 μS/cm at 20 °C (50 mg/ml water)

solubility

H2O: 50 mg/mL at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

suitability

in accordance for electrophoresis test

storage temp.

2-8°C

Other Notes

Polyacrylamide gel electrophoresis of nucleic acids[1]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roongnapa Suedee et al.
Journal of chromatography. A, 1114(2), 239-249 (2006-03-15)
A temperature sensitive dopamine-imprinted polymer was prepared in 80% aqueous methanol solution by free-radical cross-linking co-polymerisation of methacrylic acid and acrylamide at 60 degrees C in the presence of N,N-methylene-bis-acrylamide as the cross-linker and dopamine hydrochloride as template molecule. The
M Nordin-Andersson et al.
Cell biology and toxicology, 19(1), 43-51 (2003-03-29)
Basal cytotoxicity, morphological changes and alterations in cell physiological and neurochemical functions were studied in differentiated human neuroblastoma (SH-SY5Y) cells during exposure to acrylamide and during a subsequent recovery period after cessation of exposure. Acrylamide induced a 20% reduction in
R.C. Ogden et al.
Methods in Enzymology, 152, 66-66 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service