Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63132

Sigma-Aldrich

Magnesium phosphate tribasic octahydrate

purum p.a., ≥97.0% (KT)

Synonym(s):

Trimagnesium phosphate octahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg3(PO4)2 · 8H2O
CAS Number:
Molecular Weight:
406.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥97.0% (KT)
form:
powder

grade

purum p.a.

Assay

≥97.0% (KT)

form

powder

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Ca: ≤1000 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Na: ≤2000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

O.O.O.O.O.O.O.O.[Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/3Mg.2H3O4P.8H2O/c;;;2*1-5(2,3)4;;;;;;;;/h;;;2*(H3,1,2,3,4);8*1H2/q3*+2;;;;;;;;;;/p-6

InChI key

MOMDCAYSCWFERX-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The System MgO-P2O5-H2O at 25? C.
Brown PW, et al.
Journal of the American Ceramic Society. American Ceramic Society, 76(6), 1558-1562 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service