Skip to Content
MilliporeSigma
All Photos(1)

Documents

31270

Sigma-Aldrich

Potassium sulfate

puriss. p.a., ACS reagent, reag. ISO, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SO4
CAS Number:
Molecular Weight:
174.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

Agency

Ph. Eur.
USP/NF
reag. ISO

Assay

≥99%

impurities

residual solvents, complies
≤0.0005% heavy metals (as Pb)
≤0.0005% total nitrogen (N)
≤0.005% in water insoluble matters

anion traces

chloride (Cl-): ≤5 mg/kg
phosphate (PO43-): ≤10 mg/kg

cation traces

As: ≤1 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤20 mg/kg
Na: ≤50 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

suitability

in accordance for appearance of solution

SMILES string

[K+].[K+].[O-]S([O-])(=O)=O

InChI

1S/2K.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

OTYBMLCTZGSZBG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 890-890 (1994)
Minimizing adsorption of proteins on fused silica in capillary zone electrophoresis by the addition of alkali metal salts to the buffers.
Green JS and Jorgenson JW.
Journal of Chromatography A, 478, 63-70 (1989)
Synthesis of needlelike mullite particles using potassium sulfate flux.
Hashimoto S and Yamaguchi A.
J. Eur. Ceram. Soc., 20(4), 397-402 (2000)
Jinyong Yan et al.
Bioresource technology, 102(7), 4755-4758 (2011-02-01)
A dual modification procedure composed of cross-linking and protein coating with K(2)SO(4) was employed to modify Geotrichum sp. lipase for catalyzing biodiesel production from waste cooking oil. Compared to single modification of protein coating with K(2)SO(4), the dual modification of
Ashley C Parker et al.
Clinical orthopaedics and related research, 469(11), 3008-3015 (2011-05-10)
Local drug delivery has substantial potential to prevent infections compared with systemic delivery. Although calcium sulfate (CaSO(4)) has been studied for local drug delivery and two types are commercially available, it is unknown whether they differentially release antibiotics. We determined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service