Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

31173

Sigma-Aldrich

Ammonium cerium(IV) sulfate dihydrate

≥99%

Synonym(s):

Ceric ammonium sulfate, Cerium(IV) ammonium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NH4)4(SO4)4 · 2H2O
CAS Number:
Molecular Weight:
632.55
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99%

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤0.005% heavy metals (as Pb)
≤0.005% insoluble in acid

mp

130 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤50 mg/kg

cation traces

Fe: ≤50 mg/kg

SMILES string

N.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O

InChI

1S/Ce.4H3N.4H2O4S.2H2O/c;;;;;4*1-5(2,3)4;;/h;4*1H3;4*(H2,1,2,3,4);2*1H2/q+4;;;;;;;;;;/p-4

InChI key

VCNAMBGKEDPVGQ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium cerium(IV) sulfate dihydrate (tetraammonium cerium(IV) sulphate dihydrate) is commonly employed as starting material for various organic syntheses. It is widely employed as oxidizing agent. Its thermal decomposition and thermal characteristics have been reported.

Application

Ammonium cerium(IV) sulfate may be used for the determination of urinary iodine by ammonium persulfate digestion on microplate (APDM) method and colorimetric method.
Ammonium cerium(IV) sulfate was used in thin-layer chromatography for the detection of anti-depreβant drugs in human blood and urine samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Sadeg et al.
Clinical chemistry, 43(3), 498-504 (1997-03-01)
We evaluated the clinical utility of an automated HPLC system (Remedi, Bio-Rad) for identification of drugs and metabolites in biological fluids. Serum or urine or both from 354 consecutive cases of poisoning were analyzed by the system and by a
Fast colorimetric method for measuring urinary iodine.
Daniella Gnat et al.
Clinical chemistry, 49(1), 186-188 (2003-01-01)
T Ohashi et al.
Clinical chemistry, 46(4), 529-536 (2000-04-12)
Urinary iodine is a good biochemical marker for control of iodine deficiency disorders. Our aim was to develop and validate a simple, rapid, and quantitative method based on the Sandell-Kolthoff reaction, incorporating both the reaction and the digestion process into
Thermal behaviour of sulphato and nitrato complexes of cerium (IV).
Pokol G, et al.
J. Therm. Anal., 42(2-3), 343-359 (1994)
K H Gordon et al.
Organic letters, 3(1), 53-56 (2001-06-30)
[figure: see text] A novel benzyloxyaniline linker that uses ceric ammonium nitrate (CAN) as a cleavage reagent is described. Its application in the solid-phase synthesis of N-unsubstituted beta-lactams and secondary amides furnishes compounds in moderate to excellent yield (45-91%) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service