Skip to Content
MilliporeSigma
All Photos(1)

Documents

31125

Sigma-Aldrich

Barium chloride dihydrate

puriss. p.a. plus, ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaCl2 · 2H2O
CAS Number:
Molecular Weight:
244.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

ACS reagent
puriss. p.a. plus

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Assay

≥99%

form

crystals

impurities

≤0.002% total nitrogen (N)

loss

14.0-16.0% loss on drying, 150 °C

pH

5.2-8.0 (25 °C, 5%)

cation traces

Ca: ≤50 mg/kg
Fe: ≤1 mg/kg
K: ≤25 mg/kg
Mg: ≤10 mg/kg
Na: ≤50 mg/kg
Sr: ≤100 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

O.O.Cl[Ba]Cl

InChI

1S/Ba.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

PWHCIQQGOQTFAE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluating the performance of peroxide and conjugated diene values in monitoring quality of used frying oils.
Farhoosh R and Moosavi SMR.
Journal of Agriculture, Science and Technology, 11, 173-179 (2010)
Proton magnetic resonance study of the structure of barium chloride dihydrate.
Silvidi AA and McGrath JW.
J. Chem. Phys. , 32(3), 924-926 (1960)
Evaluation of quality changes in walnut kernels (Juglans regia L.) by Vis/NIR spectroscopy.
Jensen PN, et al.
Journal of Agricultural and Food Chemistry, 49(12), 5790-5796 (2001)
Barium chloride dihydrate by neutron diffraction.
Padmanabhan VM, et al.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 34(7), 2290-2292 (1978)
Eagleson M.
Concise Encyclopedia Chemistry, 116-116 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service