Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

25038

Sigma-Aldrich

Copper(II) acetate monohydrate

≥99% (iodometric)

Synonym(s):

Cupric acetate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(CO2CH3)2 · H2O
CAS Number:
Molecular Weight:
199.65
Beilstein:
3730548
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

vapor density

6.8 (vs air)

Assay

≥99% (iodometric)

reaction suitability

reagent type: catalyst
core: copper

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤5000 mg/kg

cation traces

Fe: ≤50 mg/kg
Pb: ≤20 mg/kg
Zn: ≤100 mg/kg

SMILES string

O.CC(=O)O[Cu]OC(C)=O

InChI

1S/2C2H4O2.Cu.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2

InChI key

NWFNSTOSIVLCJA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yangyang Zhao et al.
Nanoscale, 11(24), 11922-11932 (2019-06-13)
The present work demonstrates development of a miniaturized plasmonic platform comprised of a Au nanohole array (NHA) on a Si/Si3N4 substrate. Plasmonic responses of the NHA platform, which is coated with Cu-benzenetricarboxylate metal organic framework (MOF), are found to be
Marie Poulain et al.
Environmental science & technology, 53(3), 1157-1164 (2018-12-24)
Estimates of cumulative plastic inputs into the oceans are expressed in hundred million tons, whereas the total mass of microplastics afloat at sea is 3 orders of magnitude below this. This large gap is evidence of our ignorance about the
Alexander Navarrete et al.
Faraday discussions, 183, 249-259 (2015-09-24)
A novel plasmonic reactor concept is proposed and tested to work as a visible energy harvesting device while allowing reactions to transform CO2 to be carried out. Particularly the reverse water gas shift (RWGS) reaction has been tested as a
Łukasz Orzeł et al.
Dalton transactions (Cambridge, England : 2003), 44(13), 6012-6022 (2015-02-28)
The nature of chlorophyll interactions with copper(II) ions varies considerably in organic solvents, depending on the dominant coordinative form. Besides formation of the metallo tetrapyrrolic complex, Cu(II) ions can cause oxidation of the pigment, reversible or irreversible, which can lead
Jannet Kamoun et al.
Biochimica et biophysica acta, 1851(2), 129-140 (2014-12-03)
Yarrowia lipolytica is a lipolytic yeast possessing 16 paralog genes coding for lipases. Little information on these lipases has been obtained and only the major secreted lipase, namely YLLIP2, had been biochemically and structurally characterized. Another secreted lipase, YLLIP8, was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service