Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

24102

Sigma-Aldrich

Ethyl alcohol, pure

≥99.8% (GC), absolute

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
EC Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.21
technique(s):
GC/GC: suitable
bp:
78 °C (lit.)
vapor pressure:
44.6 mmHg ( 20 °C)

Product Name

Ethanol, absolute, ≥99.8% (GC)

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

grade

absolute

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

683 °F

expl. lim.

19 %, 60 °F

technique(s)

GC/GC: suitable

impurities

≤0.0005% free alkali (as NH3)
≤0.001% free acid (as CH3COOH)
≤0.005% aldehydes (as CH3CHO)
≤0.01% non-volatile matter
≤0.1% methanol (GC)
≤0.2% water (Karl Fischer)

refractive index

n20/D 1.3600 (lit.)

pH

7.0 (20 °C, 10 g/L)

bp

78 °C (lit.)

mp

−114 °C (lit.)

format

neat

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Pertains only in Germany: Nicht zu Fertigungszwecken1

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

greener alternative product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physicochemical Properties of SnO2-Sb2 O 5 Films Prepared by the Spray Pyrolysis Technique.
Correa-Lozano B, et al.
Journal of the Electrochemical Society, 143(1), 203-209 (1996)
Spectrophotometric characteristics of chlorophylls a and b and their pheophytins in ethanol.
J F Wintermans et al.
Biochimica et biophysica acta, 109(2), 448-453 (1965-11-29)
50-Hz extremely low frequency electromagnetic fields enhance cell proliferation and DNA damage: possible involvement of a redox mechanism.
Wolf FI, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1743(1), 120-129 (2005)
Fabrication of thin film dye sensitized solar cells with solar to electric power conversion efficiency over 10%.
Ito S, et al.
Thin Solid Films, 516(14), 4613-4619 (2008)
Synthesis of ZnO nanorod and the annealing effect on its photoluminescence property.
Wu L, et al.
Opt. Mater., 28(4), 418-422 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service