Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14304

Sigma-Aldrich

Sodium tungstate dihydrate

purum, ≥99%

Synonym(s):

Tungstic acid sodium salt dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2WO4 · 2H2O
CAS Number:
Molecular Weight:
329.85
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥99%
form:
crystalline

grade

purum

Assay

≥99%

form

crystalline

reaction suitability

reagent type: catalyst
core: tungsten

impurities

≤0.1% free alkali (as NaOH)

pH

9-11.5 (20 °C, 100 g/L)

mp

698 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Fe: ≤20 mg/kg
Mo: ≤100 mg/kg

SMILES string

O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O

InChI

1S/2Na.2H2O.4O.W/h;;2*1H2;;;;;/q2*+1;;;;;2*-1;

InChI key

WPZFLQRLSGVIAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Infrared absorption and differential scanning calorimetry studies of sodium tungstate dihydrate glasses containing iron oxide.
Al-Hawery AS.
Journal of Physics and Chemistry of Solids, 58(9), 1325-1328 (1997)
Luminescence and absorbance of highly crystalline CaMoO4, SrMoO4, CaWO4 and SrWO4 nanoparticles synthesized by co-precipitation method at room temperature.
Thongtem T, et al.
J. Alloy Compounds, 506(1), 475-481 (2010)
Alexander V Agafonov et al.
Molecules (Basel, Switzerland), 24(18) (2019-09-22)
The electrorheological (ER) effect was experimentally observed in dielectric suspensions containing tungsten oxide (WO3) modified with surfactant molecules (sodium dodecyl sulfate (SDS) and dodecylamine (DDA)) in electric fields up to several kilovolts per millimeter. The dielectric properties of WO3 suspensions
Marta Amigó-Correig et al.
PloS one, 7(7), e39087-e39087 (2012-07-18)
This study aims at exploring the effects of sodium tungstate treatment on hypothalamic plasticity, which is known to have an important role in the control of energy metabolism. Adult lean and high-fat diet-induced obese mice were orally treated with sodium
Andrew R Osterburg et al.
Journal of immunotoxicology, 7(3), 174-182 (2010-02-25)
The potential for adverse health effects of using tungsten and its alloys in military munitions are an important concern to both civilians and the US military. The toxicological implications of exposure to tungsten, its alloys, and the soluble tungstate (Na(2)WO(4))

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service