Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13447

Sigma-Aldrich

Sodium nitrite

puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 250, 99-100.5% (calc. to the dried substance)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaNO2
CAS Number:
Molecular Weight:
69.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form:
solid

grade

puriss.

Assay

99-100.5% (calc. to the dried substance)

form

solid

autoignition temp.

914 °F

quality

meets analytical specification of Ph. Eur., BP, USP, FCC, E 250

impurities

acidity or alcalinity, in accordance
residual solvents, complies
≤0.001% heavy metals (as Pb)

loss

≤0.2% loss on drying, 4 h (on silica gel)
≤0.25% loss on drying (on vacuum)

mp

271 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤50 mg/kg
Hg: ≤1 mg/kg
Pb: ≤4 mg/kg

suitability

complies for appearance of solution

SMILES string

[Na+].[O-]N=O

InChI

1S/HNO2.Na/c2-1-3;/h(H,2,3);/q;+1/p-1

InChI key

LPXPTNMVRIOKMN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Combined effect of NaNO2 and NaCl on the growth of Listeria monocytogenes in tryptone soya broth has been evaluated using automated turbidimetric system. Its carcinogenic effects have been examined in F-344 rats.

Application

Sodium nitrite has been used for the quantitative determination of nitrite in culture medium. It may be used in the preparation of isopropyl nitrite, via reaction with isope alcohol.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Maekawa et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 20(1), 25-33 (1982-02-01)
The carcinogenicity of sodium and of sodium nitrate was examined in F-344 rats. Sodium nitrite was administered in the drinking-water for 2 yr at levels of 0.125 or 0.25%. Sodium nitrate was given in the diet at levels 2.5 or
Oscar Zannou et al.
Molecules (Basel, Switzerland), 25(16) (2020-08-23)
Deep eutectic solvents (DESs) have got huge interest as new green and sustainable solvents for the extraction of bioactive compounds from plants in recent decades. In the present study, we aimed to investigate the effectiveness of hydrophilic DES for the
Eagleson M.
Concise Encyclopedia Chemistry, 556-556 (1994)
P J McClure et al.
International journal of food microbiology, 14(1), 77-91 (1991-10-01)
An automated turbidimetric system using multiwelled plates was used to examine the effects of different combinations of NaCl (0.5-8.0% w/v), NaNO2 (0-400 micrograms/ml) pH (4.6-7.4) and temperature (5-30 degrees C) on the growth of Listeria monocytogenes in tryptone soya broth.
R Kokkola et al.
Scandinavian journal of immunology, 61(1), 1-9 (2005-01-13)
Abstract High-mobility group box chromosomal protein 1 (HMGB1) is a protein with both intranuclear functions and extracellular cytokine-like effects. In this report, we study possible candidate receptors for HMGB1 on macrophages (Mphi) and define pathways activated by HMGB1 binding. Bone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service