Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12639

Sigma-Aldrich

Potassium hexacyanoferrate(II) trihydrate

purum, ≥99%

Synonym(s):

Potassium ferrocyanide, Yellow prussiate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K4Fe(CN)6 · 3H2O
CAS Number:
Molecular Weight:
422.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

purum

Assay

≥99%

form

crystalline

reaction suitability

reagent type: catalyst
core: iron

impurities

≤0.02% water insoluble matter

pH

8-10 (25 °C, 211 g/L)

mp

70 °C (lit.)

anion traces

chloride (Cl-): ≤5000 mg/kg
sulfate (SO42-): ≤1000 mg/kg

SMILES string

O.O.O.[K+].[K+].[K+].[K+].N#C[Fe-4](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/6CN.Fe.4K.3H2O/c6*1-2;;;;;;;;/h;;;;;;;;;;;3*1H2/q;;;;;;-4;4*+1;;;

InChI key

NRYOVBQTVIWDNU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Concha Bosch-Navarro et al.
Physical chemistry chemical physics : PCCP, 17(44), 29628-29636 (2015-10-20)
The electrochemical activity of graphene is of fundamental importance to applications from energy storage to sensing, but has proved difficult to unambiguously determine due to the challenges innate to fabricating well defined graphene electrodes free from contamination. Here, we report
Shao Su et al.
Nanoscale, 7(45), 19129-19135 (2015-11-03)
Herein, a facile approach for shape-controlled gold nanoparticle (AuNP) decorated thionine-MoS2 nanocomposites (AuNP-Thi-MoS2) had been reported by using the synergistic effect of MoS2 and Thi. Thi was not only used as an electrochemical indicator, but also as a reducing agent
Naveen Kumar et al.
Analytical biochemistry, 456, 43-49 (2014-04-15)
Here, we report a simple and sensitive colorimetric method for detection of melamine in milk using gold nanoparticles (AuNPs). AuNPs of 21-nm size were synthesized by the citrate reduction method. The method is based on the principle that the melamine
Anna Miodek et al.
Sensors (Basel, Switzerland), 15(10), 25015-25032 (2015-10-02)
An aptasensor with enhanced anti-fouling properties has been developed. As a case study, the aptasensor was designed with specificity for human thrombin. The sensing platform was developed on screen printed electrodes and is composed of a self-assembled monolayer made from
Peter S Toth et al.
The Analyst, 140(6), 1947-1954 (2015-02-11)
Two-phase voltammetry has been carried out using a reverse cell configuration, i.e. with the lower density organic solvent on the top of the aqueous solution in the cell, where the organic solvents contain either nitrile or ketone functional groups. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service