Skip to Content
MilliporeSigma
All Photos(1)

Documents

11217

Sigma-Aldrich

Ammonium nitrate

puriss., ≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4NO3
CAS Number:
Molecular Weight:
80.04
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

puriss.

Assay

≥98%

form

solid

impurities

≤0.001% heavy metals (as Pb)

pH

4.5-6.0 (20 °C, 5%)

bp

210 °C (lit.)

mp

169 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Na: ≤50 mg/kg

SMILES string

N.O[N+]([O-])=O

InChI

1S/HNO3.H3N/c2-1(3)4;/h(H,2,3,4);1H3

InChI key

PRORZGWHZXZQMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1C - Ammonium nitrate and ammonium nitrate containing preparations

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sari Palmroth et al.
Annals of botany, 111(3), 467-477 (2013-01-10)
Water and nitrogen (N) are two limiting resources for biomass production of terrestrial vegetation. Water losses in transpiration (E) can be decreased by reducing leaf stomatal conductance (g(s)) at the expense of lowering CO(2) uptake (A), resulting in increased water-use
Mohammad Mahdi Najafpour et al.
Dalton transactions (Cambridge, England : 2003), 41(34), 10156-10160 (2012-07-27)
Nanoscale manganese oxides within Faujasite zeolite have been synthesized with a simple method and characterized by scanning electron microscopy, X-ray diffraction spectrometry, N(2) adsorption-desorption isotherms, transmission electron microscopy, and atomic absorption spectroscopy. These oxides showed efficient water oxidizing activity in
Deborah Wakeham et al.
Physical chemistry chemical physics : PCCP, 14(15), 5106-5114 (2012-01-31)
The nanostructure of the ethanolammonium nitrate (EtAN)-air surface has been investigated using X-ray reflectometry (XRR), vibrational sum frequency spectroscopy (VSFS) and neutral impact collision ion scattering spectroscopy (NICISS). The XRR data decays more rapidly than expected for a perfectly sharp
Swathi A Turlapati et al.
FEMS microbiology ecology, 83(2), 478-493 (2012-09-15)
At the Harvard Forest, Petersham, MA, the impact of 20 years of annual ammonium nitrate application to the mixed hardwood stand on soil bacterial communities was studied using 16S rRNA genes pyrosequencing. Amplification of 16S rRNA genes was done using
Manuel E Ortiz-Santaliestra et al.
Aquatic toxicology (Amsterdam, Netherlands), 110-111, 170-176 (2012-02-14)
Temporary ponds where many amphibians breed experience a gradual desiccation that leads to growing larval densities, which can reduce tadpole survival rates and increase cannibalistic interactions among carnivorous salamander larvae. Concentrations of many agrochemicals, including nitrogenous fertilizers, can also increase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service