Skip to Content
MilliporeSigma
All Photos(1)

Documents

04101

Sigma-Aldrich

Polyphosphoric acid

≥83% phosphate (as P2O5) basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hn+2PnO3n+1
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38070203

Assay

≥83% phosphate (as P2O5) basis

bp

300 °C (lit.)

cation traces

As: ≤30 mg/kg
Cu: ≤1 mg/kg
Fe: ≤20 mg/kg
Pb: ≤1 mg/kg

InChI

1S/O5P2.H3O4P/c1-2-7-4-3-6(1)5-7;1-5(2,3)4/h;(H3,1,2,3,4)

InChI key

FVYIVXLIMHNYTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyphosphoric acid (PPA) is an inorganic acid. Galangin-KMnO4-PPA system has been employed for the determination of tryptophan. It participates in the Skraup reaction. It is widely employed reagent for the alkylation and acylation reactions of aromatic and alkenic compounds.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Covalent modification of vapour-grown carbon nanofibers via direct Friedel-Crafts acylation in polyphosphoric acid.
Baek J-B, et al.
Journal of Materials Chemistry, 14(13), 2052-2056 (2004)
Leyla Yurttaş et al.
Journal of enzyme inhibition and medicinal chemistry, 30(3), 458-465 (2014-09-10)
Twenty-five new N-[4-(benzothiazole-2-yl)phenyl]acetamide derivatives bearing different heterocyclic ring systems were synthesized using 2-(4-aminophenyl)benzothiazole structure as a pharmacophoric group. Final compounds were screened for their potential antitumor activity in vitro against approximately 60 human tumor cell lines derived from nine neoplastic
906. Synthetic uses of polyphosphoric acid.
Denton DA and Suschitzky H.
Journal of the Chemical Society, 4741-4743 (1963)
Li Li et al.
Luminescence : the journal of biological and chemical luminescence, 30(5), 512-518 (2014-10-02)
A novel galangin-potassium permanganate (KMnO4)-polyphosphoric acid (PPA) system was found to have an outstanding response to tryptophan (Trp). Trp determination using this KMnO4 -PPA system was enhanced significantly in the presence of galangin. A highly sensitive flow-injection chemiluminescence (CL) method
Jean-Philippe St-Pierre et al.
Acta biomaterialia, 6(8), 3302-3309 (2010-03-02)
A major challenge to the successful clinical application of bioengineered cartilage remains its integration to surrounding tissues upon implantation. One way to address this consists of generating biphasic constructs composed of articular cartilage formed in vitro on the top surface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service