Key Documents
Z805734
Specac® Omni Cell spacers
size 1.00 mm , rectangular, PTFE
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
material
PTFE
packaging
pkg of 5 ea
manufacturer/tradename
Specac Ltd
technique(s)
IR spectroscopy: suitable
size
1.00 mm , rectangular
Legal Information
Specac is a registered trademark of Specac Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)