Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z805491

FluoroSELECT single channel fluorometer

fluorescence λex 530 nm; λem 590 nm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

AC/DC input

100 V / 240 V, 50/60 Hz, European 2-pin plug (by included adaptor)
100 V / 240 V, 50/60 Hz, US 2-pin plug

usage

200 μL (mini vials or PCR tubes)

feature

CE compliant

size

185 mm × 90 mm × 35 mm

battery power

4 AA (not included)

fluorescence

λex 530 nm; λem 590 nm

Looking for similar products? Visit Product Comparison Guide

General description

The FluoroSELECT fluorometer is a portable instrument designed for multipurpose fluorescence measurements. The instrument is simple to use, light in weight, and can be powered by either DC power adaptor or battery, making it an excellent choice for field studies and laboratory measurements.

Key Features:
  • Usable with either 200 μL glass mini-tubes or PCR tubes
  • Measurements can be performed with as little as 100 μL of sample when using glass mini-tubes
  • LCD touch-screen display
  • User-friendly software with “touch and test” operation
  • USB interface for data management
  • Compact and robust
  • Larger than 6 logs of dynamic range with proper calibration procedure (assay test dependant)

Includes:
  • FluoroSELECT fluorometer
  • 5V DC power adapter (100-240 V) with 2-pin US plug and 2-pin Euro plug adaptor.
  • USB cable
  • Operation manual and USB driver/data management software CD
The FluoroSELECT product line combines portable, low cost, single-channel fluorometers with suitable kits for a wide range of applications.


Learn more about our FluoroSELECT

Legal Information

FluoroSELECT is a trademark of Sigma-Aldrich International GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service