Key Documents
Z800864
Hellma® fluorescence cuvettes, sealable cells
Suprasil® quartz, spectral range 200-2500 nm, pathlength 10x10 mm, chamber volume 3500 μL
About This Item
UNSPSC Code:
12352200
material
Suprasil® quartz
description
4 windows
manufacturer/tradename
Hellma 117.200F-QS
parameter
200-2500 nm spectral range
H × W × D
56 mm × 12.5 mm × 12.5 mm
chamber volume
3500 μL
pathlength
10x10 mm
General description
macro, semi-micro, for anaerobic applications
Legal Information
Hellma is a registered trademark of Hellma GmbH
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Choose from one of the most recent versions:
Certificates of Analysis (COA)